missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Control Cable RJ 45 for Vacuum Pumps and Controllers

Catalog No. 361019950
Change view
Click to view available options
For Use With (Equipment):
Vacuum pump and controllers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
361019950 Vacuum pump and controllers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361019950 Supplier BUCHI Supplier No. 044989

Communication cable. Connection between vacuum controller/interface and recirculating chiller or vacuum controller/interface and vacuum pump.

  • Gray Ethernet cable
  • 2m
  • Measuring range: 0-14000hPa (mbar)
  • Ambient temperature range: 10 to 40°C

Specifications

Color Gray
Type RJ45 Cat.5e Ethernet Cable
For Use With (Equipment) Vacuum pump and controllers
Length (Metric) 2 m