Learn More
BUCHI Condensers for BUCHI Rotavapors
Catalog No. 05402495
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-205, R-210/R-215
Rotavapor R-80
Material (Coating):
P+G
Plastic and Glass
Type:
Cold Trap Condenser
Condenser C
Vertical Condensor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material (Coating) Type
05402495 Rotavapor R-205, R-210/R-215 P+G Condenser C
01673000 Rotavapor R-80 Plastic and Glass Cold Trap Condenser
01672999 Rotavapor R-80 Plastic and Glass Vertical Condensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05402495 Supplier BUCHI Supplier No. 040642
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
Specifications
Material Borosilicate Glass (3.3)
Type Condenser C
For Use With (Application) Evaporator Condensation
For Use With (Equipment) Rotavapor R-205, R-210/R-215
Material (Coating) P+G
Quantity 1 Ea.