Learn More
Synthware™ Bubbler
GREENER_CHOICE
Bubbler has a small ball joint to prevent oil from being sucked back under sudden negative pressure
$216.29 - $234.89
Specifications
Material Borosilicate Glass
Type Mineral Oil Bubbler
Length (Metric) 157 mm
For Use With (Application) Organic Synthesis
Quantity 1 Each
View More Specs
Products 3
Catalog Number Mfr. No. Includes Standard Taper Joint Price Quantity & Availability  
Catalog Number Mfr. No. Includes Standard Taper Joint Price Quantity & Availability  
31500703
GREENER_CHOICE
View Documents Promotion Details
Synthware™
B262914
Ground Joint on the Bottom 29/42, 14/20
Each for $221.96
Only null left
31500704
GREENER_CHOICE
View Documents Promotion Details
Synthware™
B262924
Ground Joint on the Bottom 29/42, 24/40
Each for $234.89
Only null left
31500705
GREENER_CHOICE
View Documents Promotion Details
Synthware™
B262910
Vacuum Port on the Bottom 29/42
Each for $216.29
Only null left
Description
Description
Specifications
Specifications
Borosilicate Glass
157 mm
1 Each
Mineral Oil Bubbler
Organic Synthesis