Learn More
Chemglass Inc Bottle, Wide Mouth, Amber, Convenience Pack, 38-400 Thread Size, 120mL/4oz, PTFE Lined Green Cap
Catalog No. 50872737
Encompass
Bottle, Wide Mouth, Amber, Convenience Pack, 38-400 Thread Size, 120mL/4oz, PTFE Lined Green Cap. Amber bottles for protection of light sensitive materials. Large wide mouth opening provides for easier access of contents. Available with a choice of cap styles, PTFE lined, Polyseal, or Vinyl lined Pulp Supplied with caps attached in convenience packs.
Catalog No. 50872737 Supplier Chemglass Inc Supplier No. CG82105
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bottle, Wide Mouth, Amber, Convenience Pack, 38-400 Thread Size, 120mL/4oz, PTFE Lined Green Cap. Amber bottles for protection of light sensitive materials. Large wide mouth opening provides for easier access of contents. Available with a choice of cap styles, PTFE lined, Polyseal, or Vinyl lined Pulp Supplied with caps attached in convenience packs.