missing translation for 'onlineSavingsMsg'
Learn More

Bsu DNA Polymerase Large Fragment, Lyophilized, G-Biosciences

Catalog No. S000275
Change view
Click to view available options
Quantity:
1000 U
200 U
2000 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000275 200 U
S000265 1000 U
S000262 2000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S000275 Supplier G-Biosciences Supplier No. 7862150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bsu DNA Polymerase, large fragment is from Bacillus subtilis.

It is modified to retain its 5'-to-3' DNA polymerase activity with its 5'-to-3' exonuclease domain removed. It also lacks the 3'-to-5' exonuclease activity.

Specifications

Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method End-point PCR
Reaction Speed 20 min.
Label or Dye Biotin, FAM, TAMRA
Product Type DNA Polymerase
Quantity 200 U
For Use With (Application) Microbial Detection, Pathogen Detection, Isothermal Amplification