missing translation for 'onlineSavingsMsg'
Learn More

BSTFA 95.0+%, TCI America™

Catalog No. a56035ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56035ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56035ML Supplier TCI America Supplier No. A56035ML
Only null left

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 mL
UN Number 2924
Formula Weight 257.40
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material BSTFA [=N,O-Bis(trimethylsilyl)trifluoroacetamide] [for Gas Chromatography]
EINECSNumber (2)-3613
TSCA No
Recommended Storage Refrigerator