missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Supplier:  Thermo Scientific™ TS38833

 View more versions of this product

Catalog No. PI38833


Only null left

Description

Description

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone
Specifications

Chemical Identifiers

2°C to 8°C

Specifications

GC
BSTFA +1% TMCS; 25 g
25 g
98 to 100, 0.1 to 1%
2°C to 8°C
BSTFA +1% TMCS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.