missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Catalog No. PI38832
Encompass
Change view
Click to view available options
Standard Components:
BSTFA +1% TMCS
Quantity:
10 x 1 mL ampules
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components Quantity
PI38832 BSTFA +1% TMCS 10 g
PI38831 BSTFA +1% TMCS 10 x 1 mL ampules
PI38833 BSTFA +1% TMCS 25 g
PI38834 BSTFA +1% TMCS 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI38832 Supplier Thermo Scientific™ Supplier No. TS38832

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone

Specifications

For Use With GC
Standard Concentration 98 to 100, 0.1 to 1%
Storage 2°C to 8°C
Quantity 10 g
Standard Components BSTFA +1% TMCS

For Research Use Only. Not for use in diagnostic procedures.