Learn More
New England Biolab Ltd Bst 3.0 DNA Polymerase 1600 units
Catalog No. 501047481
Encompass
Bst 3.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment engineered for improved isothermal amplification performance and increased reverse transcription activity. Bst 3.0 DNA Polymerase contains 5' to 3' DNA polymerase activity with either DNA or RNA templates and strong strand displacement activity, but lacks 5' to 3' and 3' to 5' exonuclease activity. Bst 3.0 DNA Polymerase demonstrates robust performance even in high concentrations of amplification inhibitors and features significantly increased reverse transcriptase activity compared to Bst DNA Polymerase.
Catalog No. 501047481 Supplier New England Biolab Ltd Supplier No. M0374S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bst 3.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment engineered for improved isothermal amplification performance and increased reverse transcription activity. Bst 3.0 DNA Polymerase contains 5' to 3' DNA polymerase activity with either DNA or RNA templates and strong strand displacement activity, but lacks 5' to 3' and 3' to 5' exonuclease activity. Bst 3.0 DNA Polymerase demonstrates robust performance even in high concentrations of amplification inhibitors and features significantly increased reverse transcriptase activity compared to Bst DNA Polymerase.