Learn More
New England Biolab Ltd Bst 2.0 DNA Polymerase 1600 units
Catalog No. NC0299281
Encompass
Bst 2.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment). Bst 2.0 DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand displacement activity but lacks 5' to 3' exonuclease activity. Bst 2.0 DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.
Catalog No. NC0299281 Supplier New England Biolab Ltd Supplier No. M0537S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bst 2.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment). Bst 2.0 DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand displacement activity but lacks 5' to 3' exonuclease activity. Bst 2.0 DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.