missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTRAX™ Brush Step™ Ribbed Surface Mat
Description
- Ribbed surface brushes, channels, and conceals dirt and debris
- Ribs run length of mat
- Ideal choice for light to medium traffic areas
- 18 ounces of needle-punched yarn per square yard
- Low profile to perform in narrow clearance doorways
- Vinyl backing ensures minimal buckling and mat movement.
Specifications
Specifications
| Surface | Ribbed surface |
| Dimensions (L x W) | 4 x 6 ft. |
| Width (English) | 6 ft. |
| Quantity | 1 Ea. |
| Color | Charcoal |
| Length (English) | 4 ft. |