missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bronze gauze, alloy 220, 200 mesh woven from 0.053mm (0.0021in) in dia wire, Thermo Scientific™
Specifications
| Chemical Name or Material | Bronze gauze, Alloy 220 |
|---|---|
| Name Note | 200 mesh woven from 0.053mm (0.0021 in.) dia. wire |
| MDL Number | MFCD00198187 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA45223GT
|
Thermo Scientific Chemicals
45223GT |
150 x 150 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AA45223HB
|
Thermo Scientific Chemicals
45223HB |
300 x 300 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Bronze gauze, Alloy 220 | |
| 200 mesh woven from 0.053mm (0.0021 in.) dia. wire | |
| MFCD00198187 |