missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bronze Gauze, 20 mesh woven from 0.41mm (0.016 in.) in dia. wire, Thermo Scientific™
$57.11 - $57.11
Spécifications
| Chemical Name or Material | Bronze |
|---|---|
| Appearance | Bronze colored gauze |
| Mesh Size | 220,20 Mesh |
| Odor | Odorless |
| Form | Gauze |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA45144GT
|
Thermo Scientific Chemicals
45144GT |
150 x 150 mm |
chaque for $57.11
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| Bronze | |
| 220,20 Mesh | |
| Gauze |
| Bronze colored gauze | |
| Odorless | |
| MFCD00198187 |