missing translation for 'onlineSavingsMsg'
Learn More

Brompheniramine maleate

Catalog No. AAJ63922MB
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
AAJ63922MB 25 mg
1 options

Catalog No. AAJ63922MB

Supplier: Thermo Scientific Chemicals J63922MB

Only null left

An antihistamine and also an inhibitor of serotonin reuptake | CAS: 980-71-2 | C20H23BrN2O4 | 435.32 g/mol

An antihistamine and also an inhibitor of serotonin reuptake

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 980-71-2
Color White
Molecular Formula C20H23BrN2O4
Quantity 25 mg
MDL Number MFCD00057367
Merck Index 14,1443
Synonym brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
InChI Key SRGKFVAASLQVBO-BTJKTKAUNA-N
SMILES OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1
IUPAC Name (2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
Molecular Weight (g/mol) 435.32
PubChem CID 5281067
ChEBI CHEBI:3184
Formula Weight 319.24
Physical Form Powder
Chemical Name or Material Brompheniramine maleate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-562-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only