missing translation for 'onlineSavingsMsg'
Learn More

Brompheniramine Maleate 98.0+%, TCI America™

Supplier:  TCI America B516725G

Catalog No. B516725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Brompheniramine Maleate
136°C
C20H23BrN2O4
25 g
SRGKFVAASLQVBO-BTJKTKAUNA-N
(2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
5281067
435.32
Crystalline Powder
980-71-2
White
MFCD00057367
brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1
435.32
CHEBI:3184
≥98.0% (HPLC,T)

Chemical Identifiers

980-71-2
435.32
SRGKFVAASLQVBO-BTJKTKAUNA-N
5281067
(2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
C20H23BrN2O4
MFCD00057367
brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
CHEBI:3184
OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : US4025000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions