missing translation for 'onlineSavingsMsg'
Learn More

Bromoxylenol Blue, TCI America™

Catalog No. B09985G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09985G Supplier TCI America Supplier No. B09985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40070-59-5
Molecular Formula C23H24Br2O5S
Molecular Weight (g/mol) 572.31
MDL Number MFCD00005873
InChI Key JHWIBPVPGLHJHA-RMFGFXDFSA-N
Synonym bromoxylenol blue, 3,3-dibromo-m-xylenol sulfonephthaleun, 3',3-dibromo-p-xylenolsulfonphthalein, 3',3-dibromo-p-xylenolsulfonephthalein, bromoxylenol blue, indicator grade, dye content 95 %, 2-bromo-4-3-3-bromo-4-hydroxy-2,5-dimethylphenyl-1,1-dioxo-2,1, 3,3-bis 3-bromo-4-hydroxy-2,5-dimethylphenyl-2,1$l^ 6-benzoxathiole-1,1-dione, 3,3-bis 3-bromo-4-hydroxy-2,5-dimethylphenyl-2,1??-benzoxathiole-1,1-dione, 3,3-bis 3-bromo-4-hydroxy-2,5-dimethylphenyl benzo c 1,2-oxathiolene-1,1-dione
PubChem CID 417069
IUPAC Name (3S)-3-(3-bromo-4-hydroxy-2,5-dimethylphenyl)-3-[(1S,4S,5R,6S)-5-bromo-4-hydroxy-3,6-dimethylcyclohex-2-en-1-yl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES C[C@@H]1[C@@H](Br)[C@@H](O)C(C)=C[C@@H]1[C@]1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1C

Specifications

Melting Point 218°C
Quantity 5 g
Formula Weight 568.28
Physical Form Crystalline Powder
Chemical Name or Material Bromoxylenol Blue