missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotris(dimethylamino)phosphonium Hexafluorophosphate 98.0+%, TCI America™
$38.51 - $148.25
Chemical Identifiers
| CAS | 50296-37-2 |
|---|---|
| Molecular Formula | C6H18BrF6N3P2 |
| Molecular Weight (g/mol) | 388.07 |
| MDL Number | MFCD00191864 |
| InChI Key | XELPBWPBGHCIKX-UHFFFAOYSA-N |
| Synonym | bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate |
| PubChem CID | 10221847 |
| IUPAC Name | bromotris(dimethylamino)phosphanium; hexafluoro-λâµ-phosphanuide |
| SMILES | F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C |
Chemical Identifiers
| 50296-37-2 | |
| 388.07 | |
| XELPBWPBGHCIKX-UHFFFAOYSA-N | |
| 10221847 | |
| F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C |
| C6H18BrF6N3P2 | |
| MFCD00191864 | |
| bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate | |
| bromotris(dimethylamino)phosphanium; hexafluoro-λâµ-phosphanuide |
Specifications
| 50296-37-2 | |
| C6H18BrF6N3P2 | |
| 1 g | |
| bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate | |
| F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C | |
| 388.07 | |
| 388.07 | |
| Crystalline Powder |
| White | |
| MFCD00191864 | |
| 1759 | |
| XELPBWPBGHCIKX-UHFFFAOYSA-N | |
| bromotris(dimethylamino)phosphanium; hexafluoro-λâµ-phosphanuide | |
| 10221847 | |
| ≥98.0% (HPLC,T) | |
| Bromotris(dimethylamino)phosphonium Hexafluorophosphate |
Safety and Handling
TSCA : No