missing translation for 'onlineSavingsMsg'
Learn More

Bromotris(dimethylamino)phosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B38381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38381G 1 g
B38385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38381G Supplier TCI America Supplier No. B38381G
Only null left

Chemical Identifiers

CAS 50296-37-2
Molecular Formula C6H18BrF6N3P2
Molecular Weight (g/mol) 388.07
MDL Number MFCD00191864
InChI Key XELPBWPBGHCIKX-UHFFFAOYSA-N
Synonym bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate
PubChem CID 10221847
IUPAC Name bromotris(dimethylamino)phosphanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 388.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bromotris(dimethylamino)phosphonium Hexafluorophosphate