missing translation for 'onlineSavingsMsg'
Learn More

Bromotripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7119713
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26805G 5 g
B268025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26805G Supplier TCI America Supplier No. B26805G
Only null left

Chemical Identifiers

CAS 132705-51-2
Molecular Formula C12H24BrF6N3P2
Molecular Weight (g/mol) 466.187
MDL Number MFCD00077412
InChI Key CYKRMWNZYOIJCH-UHFFFAOYSA-N
Synonym pybrop, bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, bromotripyrrolidinophosphonium hexafluorophosphate, bromotri pyrrolidin-1-yl phosphonium hexafluorophosphate v, pybrop r, bromotri 1-pyrrolidinyl phosphonium hexafluorophosphate, bromotri pyrrolidino phosphonium hexafluorophosphate, bromo-tris-pyrrolidinophosphoniumhexafluorophosphate, bromo-tris-pyrrolidino phosphoniumhexafluorophosphate, bromo-tris-pyrrolidino-phosphoniumhexafluorophosphate
PubChem CID 2733179
IUPAC Name bromo(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Br.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 5 g
Formula Weight 466.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bromotripyrrolidinophosphonium Hexafluorophosphate
TSCA No
Recommended Storage Freezer