missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotrimethylsilane, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Silylation reagent
Specifications
Specifications
| Melting Point | -43.0°C |
| Color | Colorless to Yellow |
| Density | 1.1600g/mL |
| Boiling Point | 79.0°C |
| Flash Point | 32°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (Argentometry) |
| Packaging | Glass bottle |
| Refractive Index | 1.4200 to 1.4250 |
| Linear Formula | (CH3)3SiBr |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| EINECSNumber | 220-672- |
RUO – Research Use Only