missing translation for 'onlineSavingsMsg'
Learn More

Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 97%

Catalog No. p-7047523
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1938403 1 g
AAL1938406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1938403 Supplier Thermo Scientific Chemicals Supplier No. L1938403
Only null left

CAS: 132705-51-2 | C12H24BrF6N3P2 | 466.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132705-51-2
Molecular Formula C12H24BrF6N3P2
Molecular Weight (g/mol) 466.187
MDL Number MFCD00077412
InChI Key CYKRMWNZYOIJCH-UHFFFAOYSA-N
Synonym pybrop, bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, bromotripyrrolidinophosphonium hexafluorophosphate, bromotri pyrrolidin-1-yl phosphonium hexafluorophosphate v, pybrop r, bromotri 1-pyrrolidinyl phosphonium hexafluorophosphate, bromotri pyrrolidino phosphonium hexafluorophosphate, bromo-tris-pyrrolidinophosphoniumhexafluorophosphate, bromo-tris-pyrrolidino phosphoniumhexafluorophosphate, bromo-tris-pyrrolidino-phosphoniumhexafluorophosphate
PubChem CID 2733179
IUPAC Name bromo(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Br.F[P-](F)(F)(F)(F)F

Specifications

Quantity 1 g
Beilstein 7845702
Sensitivity Moisture sensitive
Formula Weight 466.19
Percent Purity 97%
Chemical Name or Material Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only