missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, sodium salt, pure, water soluble, indicator

Catalog No. p-77481
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151370250 25 g Glass bottle
AC151371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151370250 Supplier Thermo Scientific Chemicals Supplier No. 151370250

CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
PubChem CID 102183223
IUPAC Name sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Visual Transition Interval from pH 6.0 (yellow) to pH 7.6 (blue)
Merck Index 15, 1455
Color Brown/Orange to Green
Solubility Information Solubility in water: soluble.
Quantity 25 g
Physical Form Powder
Formula Weight 646.35
Grade Pure
Chemical Name or Material Bromothymol Blue, sodium salt
Show More Show Less
EINECSNumber 252-169-7
TSCA TSCA

RUO – Research Use Only