missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromothymol Blue, sodium salt, pure, water soluble, indicator
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| UV Spectrometry | Complies |
| Visual Transition Interval | from pH 6.0 (yellow) to pH 7.6 (blue) |
| Merck Index | 15, 1455 |
| Color | Brown/Orange to Green |
| Solubility Information | Solubility in water: soluble. |
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 646.35 |
| Show More |
Safety and Handling
| EINECSNumber | 252-169-7 |
| TSCA | TSCA |
RUO – Research Use Only