missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue Sodium Salt, ACS Grade, LabChem™
Specifications
Specifications
| CAS Max % | 100 |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Linear Formula | C27H27Br2O5S·Na |
| Decomposition Information | Sulfur compounds; Carbon monoxide; Carbon dioxide; Hydrogen bromide |
| Color | Gray/Orange |
| Solubility Information | Soluble in water |
| Quantity | 10 g |
| Physical Form | Powder |
| Formula Weight | 646.37 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |