missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue-Phenol Red Mixed Indicator, Ricca Chemical
$72.78 - $258.44
Specifications
| Chemical Name or Material | Bromothymol Blue-Phenol Red Mixed Indicator |
|---|---|
| CAS | 7732-18-5,34722-90-2,34487-61-1 |
| CAS Min % | 98.0,0.02,0.02 |
| CAS Max % | 100.0,0.03,0.02 |
| Grade | Indicator |
Specifications
| Bromothymol Blue-Phenol Red Mixed Indicator | |
| 98.0,0.02,0.02 | |
| Indicator | |
| Dark Blue | |
| Liquid |
| 7732-18-5,34722-90-2,34487-61-1 | |
| 100.0,0.03,0.02 | |
| Natural Poly Bottle | |
| 7.45 to 7.55 |