missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue-Phenol Red, Mixed Indicator, Certified, LabChem™
Supplier: LabChem LC120807
Specifications
| Bromothymol Blue-Phenol Red | |
| 99.95,0.03,0.02 | |
| C27H27Br2O5S·Na | |
| Passes Test | |
| Certified | |
| 1g/mL | |
| 1g/mL | |
| 125 mL |
| Mixed Indicator | |
| 34722-90-2,34487-61-1 | |
| Soluble in water | |
| 646.37 | |
| Poly Bottle | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
| Green | |
| Liquid |