missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue, ACS Reagent (Sultone) not water soluble, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue, ACS Reagent (Sultone) not water soluble |
| CAS | 76-59-5 |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| Molecular Formula | C27H28Br2O5S |
| Color | White |
| Synonym | Dibromothymolsulfophthalein, Bromthymol Blue |
| Quantity | 1 g |
| Physical Form | Solid |
| Molecular Weight (g/mol) | 624.3812 |
| Show More |