missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Red 80.0+%, TCI America™
$115.57 - $1427.87
Chemical Identifiers
| CAS | 2800-80-8 |
|---|---|
| Molecular Formula | C19H12Br2O5S |
| Molecular Weight (g/mol) | 512.168 |
| MDL Number | MFCD00023014 |
| InChI Key | OYCLSQDXZMROJK-UHFFFAOYSA-N |
| PubChem CID | 76047 |
| IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br |
Chemical Identifiers
| 2800-80-8 | |
| 512.168 | |
| OYCLSQDXZMROJK-UHFFFAOYSA-N | |
| 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| C19H12Br2O5S | |
| MFCD00023014 | |
| 76047 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br |
Specifications
| 2800-80-8 | |
| MFCD00023014 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br | |
| 512.168 | |
| 512.17 | |
| 1 g | |
| Bromophenol Red |
| C19H12Br2O5S | |
| OYCLSQDXZMROJK-UHFFFAOYSA-N | |
| 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 76047 | |
| ≥80.0% (HPLC) | |
| Crystal/Powder |
Safety and Handling
TSCA : No