missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Red 80.0+%, TCI America™

Catalog No. B063225G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B063225G 25 g
B06321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B063225G Supplier TCI America Supplier No. B063225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2800-80-8
Molecular Formula C19H12Br2O5S
Molecular Weight (g/mol) 512.168
MDL Number MFCD00023014
InChI Key OYCLSQDXZMROJK-UHFFFAOYSA-N
PubChem CID 76047
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 512.17
Percent Purity ≥80.0% (HPLC)
Chemical Name or Material Bromophenol Red