missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue Sodium Salt Indicator, Honeywell Fluka™
Indicator
$1203.84 - $1203.84
Chemical Identifiers
| CAS | 34725-61-6 |
|---|---|
| Molecular Formula | C19H9Br4NaO5S |
| MDL Number | MFCD00013793 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 34725-61-6 | |
| C19H9Br4NaO5S | |
| MFCD00013793 |
Specifications
| 34725-61-6 | |
| MFCD00013793 | |
| 6995701 | |
| 691.94g/mol | |
| Blue | |
| 50 g | |
| Bromophenol Blue Sodium Salt |
| C19H9Br4NaO5S | |
| NONH for all modes of transport | |
| Soluble in water | |
| Glass Bottle | |
| 4.6 | |
| Powder |
Safety and Handling
EINECSNumber : 252-170-2