missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue Sodium Salt [for Electrophoresis], TCI America™

Catalog No. B31951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31951G Supplier TCI America Supplier No. B31951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H10Br4O5S
MDL Number MFCD00013793
Synonym Sodium Bromophenol Blue, BPB

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 691.94
Physical Form Crystalline Powder
Chemical Name or Material Bromophenol Blue Sodium Salt [for Electrophoresis]