missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | >300.0°C |
| Packaging | Glass bottle |
| Beilstein | 19, I, 649 |
| Quantity | 50 g |
| SMILES | C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] |
| Formula Weight | 691.94 |
| Grade | ACS Reagent |
| Chemical Name or Material | Bromophenol Blue, sodium salt, Pure |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
RUO – Research Use Only