missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS

CAS: 34725-61-6

Supplier:  Thermo Scientific Chemicals 03263914

 View more versions of this product

Catalog No. AA3263914


Only null left
Explore available promotions

Description

Description

Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bromophenol Blue sodium salt
34725-61-6
14,1444
691.96
25 g
>300°C
MFCD00013793
Soluble in water.
ACS
Crystalline

Chemical Identifiers

34725-61-6
MFCD00013793
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 252-170-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only