missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS
Description
Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C |
| Merck Index | 14,1444 |
| Solubility Information | Soluble in water. |
| Quantity | 100 g |
| Physical Form | Crystalline |
| Formula Weight | 691.96 |
| Grade | ACS |
| Chemical Name or Material | Bromophenol Blue sodium salt |
Safety and Handling
| EINECSNumber | 252-170-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only