missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt

CAS: 34725-61-6 | C19H9Br4NaO5S | 691.96 g/mol

$81.37 - $238.32

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.96
MDL Number MFCD00013793
Synonym Bromphenol Blue, water soluble
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1689909
View Documents
Thermo Scientific Chemicals
A1689909
10 g
Each for $81.37
Only null left
 
AAA1689918
View Documents
Thermo Scientific Chemicals
A1689918
50 g
Each for $238.32
Only null left
 
Description

Description

Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.

Solubility
Soluble in water and slightly soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

34725-61-6
691.96
Bromphenol Blue, water soluble
C19H9Br4NaO5S
MFCD00013793

Specifications

>300°C
C19H9Br4NaO5S
141444
Soluble in water and slightly soluble in alcohol.
691.96
Bromophenol Blue sodium salt
34725-61-6
MFCD00013793
Bromphenol Blue, water soluble
691.96
10 g
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 252-170-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only