Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt
Description
Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.
Solubility
Soluble in water and slightly soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | >300°C |
| Merck Index | 141444 |
| Solubility Information | Soluble in water and slightly soluble in alcohol. |
| Quantity | 50 g |
| Formula Weight | 691.96 |
| Chemical Name or Material | Bromophenol Blue sodium salt |
Safety and Handling
| EINECSNumber | 252-170-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only