missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046153
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046153 5 g
60046152 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046153 Supplier Solstice Supplier No. 327125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 204°C
Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 61698
Color Red
pH 2.8 to 4.4
Quantity 5 g
Physical Form Powder
Formula Weight 669.96g/mol
Chemical Name or Material Bromophenol Blue, Reag. Ph. Eur.
EINECSNumber 204-086-2
RTECSNumber SJ7453000