missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, pure, indicator

Catalog No. AC151340250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151340250 25 g Glass bottle
AC151341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151340250 Supplier Thermo Scientific Chemicals Supplier No. 151340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Bromophenol Blue, C19H10Br4O5S, CAS Number-115-39-9, 3', 3″, 5', 3',3″,5',5″-Tetrabromophenolsulfonephthalein, BPB, 25g, 19, I, 649, CHEBI:59424, Orange-Pink to Red, 669.96, Pure, UDSAIICHUKSCKT-UHFFFAOYSA-N, Authentic | CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 279.0°C
Beilstein 19, I, 649
Merck Index 15, 1454
Solubility Information Solubility in water: 0.4g/100mL
Infrared Spectrum Authentic
Formula Weight 669.96
Grade Pure
Packaging Glass bottle
Specific Extinction 1100 min. (592nm, in buffer)
Visual Transition Interval from pH 3.0 (yellow) to pH 4.6 (blue)
Color Orange/Pink to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue, Indicator
Show More Show Less

RUO – Research Use Only