missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue Indicator, 0.04% (w/v), Ricca Chemical

Catalog No. p-3251398
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
13504 120 mL Natural Poly Bottle
R1350000250 250 mL Natural Poly Bottle
135016 500 mL Natural Poly Bottle
135032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13504 Supplier Ricca Chemical Company Supplier No. 13504
Only null left

Specifications

Chemical Name or Material Bromophenol Blue Indicator
CAS 62625-28-9, 7732-18-5
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Concentration 0.04% (w/v)
Percent Purity 0.04% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Reddish Purple
pH 4
Quantity 120 mL
Physical Form Liquid
Show More Show Less