missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, ACS Reagent (Sultone) not water soluble, Reagents
Specifications
Specifications
| Chemical Name or Material | Bromophenol Blue, ACS Reagent (Sultone) not water soluble |
| CAS | 115-39-9 |
| Molecular Formula | C19H10Br4O5S |
| Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
| Molecular Weight (g/mol) | 669.9607 |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 0.954 |
| Color | Orange |
| Quantity | 10 g |
| Show More |