missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue

Catalog No. AAA1846909
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1846909 10 g
AAA1846918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAA1846909 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' A1846909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

It is employed as an electrophoresis tracking dye and also finds its uses as an industrial dye, a laboratory acid-base indicator and a biological stain to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues. In gel electrophoresis it can be used as a color marker and as a biological stain it can be used to stain proteins and nucleic acids. Bromophenol blue is an intermediate and is used as an acid-based indicator in the pH range of 3-4.6 where the color changes from yellow to blue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an electrophoresis tracking dye and also finds its uses as an industrial dye, a laboratory acid-base indicator and a biological stain to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues. In gel electrophoresis it can be used as a color marker and as a biological stain it can be used to stain proteins and nucleic acids. Bromophenol blue is an intermediate and is used as an acid-based indicator in the pH range of 3-4.6 where the color changes from yellow to blue.

Solubility
Freely soluble in NaOH. More soluble in methyl and ethyl alcohols ,benzene, and in acetic acid. Slightly soluble in water (0.4g/100g) at 20°C. Soluble in methanol (10 mg/mL.).

Notes
Transition interval: pH 3.0 (yellow) to pH 4.6 (blue)

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym Bromphenol Blue
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point ∼270°C (decomposition)
Beilstein 61698
Merck Index 14,1444
Solubility Information Freely soluble in NaOH. More soluble in methyl and ethyl alcohols ,benzene,and in acetic acid. Slightly soluble in water (0.4g/100g) at 20°C. Soluble in methanol (10mg/mL.).
Formula Weight 669.98
Odor Odorless
Quantity 10 g
Chemical Name or Material Bromophenol Blue
missing translation for 'healthHazard1' H500
missing translation for 'healthHazard3' P210-P241
EINECSNumber 204-086-2
RTECSNumber SJ7453000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only