missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromomethyl Methyl Ether 95.0+%, TCI America™
$55.92 - $167.76
Chemical Identifiers
| CAS | 13057-17-5 |
|---|---|
| Molecular Formula | C2H5BrO |
| Molecular Weight (g/mol) | 124.965 |
| MDL Number | MFCD00000171 |
| InChI Key | JAMFGQBENKSWOF-UHFFFAOYSA-N |
| Synonym | bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether |
| PubChem CID | 83093 |
| IUPAC Name | bromo(methoxy)methane |
| SMILES | COCBr |
Chemical Identifiers
| 13057-17-5 | |
| 124.965 | |
| JAMFGQBENKSWOF-UHFFFAOYSA-N | |
| 83093 | |
| COCBr |
| C2H5BrO | |
| MFCD00000171 | |
| bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether | |
| bromo(methoxy)methane |
Specifications
| 13057-17-5 | |
| 88°C | |
| MFCD00000171 | |
| 1992 | |
| JAMFGQBENKSWOF-UHFFFAOYSA-N | |
| bromo(methoxy)methane | |
| 83093 | |
| ≥95.0% (GC) | |
| Bromomethyl Methyl Ether |
| Yellow | |
| C2H5BrO | |
| 5 g | |
| bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether | |
| COCBr | |
| 124.965 | |
| 124.97 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator