missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol purple, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219508025

 View more versions of this product

Catalog No. ICN19508025


Only null left

Description

Description

Specifications

Specifications

Bromocresol purple
C21H16Br2O5S
5', 5”-dibromo-o-cresolsulfonephthalein
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
540.22
CHEBI:86154
Brown to Purple
Crystalline Powder
115-40-2
MFCD00011681
ABIUHPWEYMSGSR-UHFFFAOYSA-N
3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
8273
540.24
25 g

Chemical Identifiers

115-40-2
540.22
ABIUHPWEYMSGSR-UHFFFAOYSA-N
8273
3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
C21H16Br2O5S
MFCD00011681
5', 5”-dibromo-o-cresolsulfonephthalein
CHEBI:86154
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions