missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple, BAKER™, J.T. Baker™
A grade of chemicals for laboratory use
$411.35 - $411.35
Chemical Identifiers
| CAS | 115-40-2 |
|---|---|
| Molecular Formula | C21H16Br2O5S |
| Molecular Weight (g/mol) | 540.22 |
| MDL Number | MFCD00011681 |
| InChI Key | ABIUHPWEYMSGSR-UHFFFAOYSA-N |
| PubChem CID | 8273 |
| ChEBI | CHEBI:86154 |
| IUPAC Name | 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| SMILES | CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
Chemical Identifiers
| 115-40-2 | |
| 540.22 | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| CHEBI:86154 | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
| C21H16Br2O5S | |
| MFCD00011681 | |
| 8273 | |
| 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
Specifications
| 115-40-2 | |
| MFCD00011681 | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 | |
| 540.22 | |
| CHEBI:86154 | |
| Wide Mouth Amber Glass Bottle | |
| Bromocresol Purple |
| C21H16Br2O5S | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
| 8273 | |
| 540.22 | |
| 10 g |