missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple, ACS Reagent (Sultone) not water soluble, Reagents
Bromocresol Purple, ACS Reagent (Sultone) not water soluble, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$121.21 - $363.54
Chemical Identifiers
| CAS | 115-40-2 |
|---|---|
| Molecular Formula | C21H16Br2O5S |
| Molecular Weight (g/mol) | 540.22 |
| InChI Key | ABIUHPWEYMSGSR-UHFFFAOYSA-N |
| Synonym | Bromcresol purple |
| IUPAC Name | 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| SMILES | CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
01-336-779
|
Labchem Inc
C61060001C2 |
1 g |
Each for $121.21
|
|
|||||
|
01-336-778
|
Labchem Inc
C61060005C2 |
5 g |
Each for $363.54
|
|
|||||
Chemical Identifiers
| 115-40-2 | |
| 540.22 | |
| Bromcresol purple | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
| C21H16Br2O5S | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
Specifications
| 115-40-2 | |
| Bromcresol purple | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 | |
| 540.22 | |
| Glass Bottle | |
| Tan | |
| Solid |
| C21H16Br2O5S | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
| Reagent | |
| 0.991 | |
| 1 g | |
| Bromocresol Purple, ACS Reagent (Sultone) not water soluble |