missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Sodium Salt, ACS Reagent Grade, Ricca Chemical

$472.60 - $472.60

Chemical Identifiers

CAS 62625-32-5
Molecular Weight (g/mol) 720 g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RDCB060010F
View Documents
Ricca Chemical Company
RDCB060010F1
10 g Pouch
Each for $472.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-32-5
720 g/mol

Specifications

62625-32-5
100.0
ACS
Dark Green
Granules
98.0
720 g/mol
Pouch
10 g
Bromocresol Green, Sodium Salt
Videos
SDS
Documents

Documents