missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green sodium salt, ACS

CAS: 62625-32-5 | C21H13Br4NaO5S

$78.34 - $271.59

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
MDL Number MFCD00148898
Synonym Bromocresol Green, water soluble
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6428406
View Documents
Thermo Scientific Chemicals
J6428406
5 g
Each for $78.34
Only null left
 
AAJ6428414
View Documents
Thermo Scientific Chemicals
J6428414
25 g
Each for $271.59
Only null left
 
Description

Description

Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

Solubility
40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

62625-32-5
C21H13Br4NaO5S
MFCD00148898
Bromocresol Green, water soluble

Specifications

230°C (decomposition)
C21H13Br4NaO5S
14,1386
40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.
ACS
5 g
Bromocresol Green sodium salt
62625-32-5
MFCD00148898
Bromocresol Green, water soluble
720.02
Brown to Red
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 263-657-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only