missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromocresol Green sodium salt, 0.04% w/v aq. soln.
CAS: 62625-32-5
$56.74 - $102.01
Chemical Identifiers
| CAS | 62625-32-5 |
|---|---|
| MDL Number | MFCD00148898 |
| Synonym | Bromocresol Green, water soluble |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 62625-32-5 | |
| MFCD00148898 | |
| Bromocresol Green, water soluble |
Specifications
| 62625-32-5 | |
| MFCD00148898 | |
| 720.01 | |
| 100 mL | |
| Bromocresol Green sodium salt |
| 0.04% w/v aq. soln. | |
| Bromocresol Green, water soluble | |
| Green | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only