missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green-Methyl Red Mixed Indicator, Ricca Chemical
$79.78 - $391.19
Specifications
| Chemical Name or Material | Bromocresol Green-Methyl Red Mixed Indicator |
|---|---|
| CAS | 7732-18-5,62625-32-5,845-10-3 |
| CAS Min % | 98.0,0.1,0.02 |
| CAS Max % | 100.0,0.1,0.02 |
| Grade | Indicator |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R121500060A
|
Ricca Chemical Company
R121500060A |
60 mL | Natural Poly Bottle |
Each for $79.78
|
|
||||
|
1215-4
|
Ricca Chemical Company
12154 |
120 mL | Natural Poly Bottle |
Each for $104.34
|
|
||||
|
1215-16
|
Ricca Chemical Company
121516 |
500 mL | Natural Poly Bottle |
Each for $157.44
|
|
||||
|
1215-32
|
Ricca Chemical Company
121532 |
1 L | Natural Poly Bottle |
Each for $391.19
|
|
||||
Specifications
| Bromocresol Green-Methyl Red Mixed Indicator | |
| 98.0,0.1,0.02 | |
| Indicator | |
| Aqueous Solution | |
| Liquid |
| 7732-18-5,62625-32-5,845-10-3 | |
| 100.0,0.1,0.02 | |
| Natural Poly Bottle | |
| Dark Brown |