missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green-Methyl Red Mixed Indicator, Alcoholic, Ricca Chemical

$163.47 - $251.20

Specifications

Chemical Name or Material Bromocresol Green-Methyl Red Mixed Indicator
CAS 7732-18-5,67-63-0,62625-32-5,845-10-3
CAS Min % 55.0,42.91,0.11,0.02
CAS Max % 57.0,44.66,0.11,0.02
Grade Indicator
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1220-4
View Documents
Ricca Chemical Company
RI12204
120 mL Natural Poly Bottle N/A
Only null left
 
1220-16
View Documents Promotion Details
Ricca Chemical Company
122016
500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Specifications

Bromocresol Green-Methyl Red Mixed Indicator
55.0,42.91,0.11,0.02
Indicator
Alcoholic Solution
Liquid
7732-18-5,67-63-0,62625-32-5,845-10-3
57.0,44.66,0.11,0.02
Natural Poly Bottle
Dark Brown
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'